Learn More
Thermo Scientific™ Oxoid™ STAA Selective Supplement
Catalog No. OXSR0151E
Change view
Click to view available options
Quantity:
10/Pk.
Catalog No. Quantity
OXSR0151E 10/Pk.
1 options
Catalog No. OXSR0151E Supplier Thermo Scientific™ Supplier No. SR0151E
Only null left
The Oxoid STAA Selective Supplement is used for the isolation of Brochothrix thermosphacta from food samples.
Oxoid STAA Selective Supplement is used for the isolation of Brochothrix thermosphacta from food samples.
  • Add to STAA Agar Base, Part No. CM0881B
  • Each vial supplements 500mL of medium
Specifications
Description STAA Selective Supplement
Form Freeze-dried
Product Type Selective Supplement
Quantity 10/Pk.
Yield Each vial for 500mL medium