Learn More
Thermo Scientific™ Oxoid™ STA Selective Supplement
Numéro de catalogue. OXSR0162E
Modifier l'affichage
Click to view available options
Quantity:
10/Pk.
Numéro de catalogue. Quantity
OXSR0162E 10/Pk.
1 options
Numéro de catalogue. OXSR0162E Fournisseur Thermo Scientific™ Code fournisseur SR0162E
Il en reste null
The Oxoid STA Selective Supplement (Supplement without actidione) is used for the isolation of Brochothrix thermosphacta from food samples.
Oxoid STA Selective Supplement (Supplement without actidione) is used for the isolation of Brochothrix thermosphacta from food samples.
  • Add to STAA Agar Base, Part No. CM0881B
  • Each vial supplements 500mL of medium
Spécifications
Description STA Selective Supplement
Form Freeze-dried
Product Type Selective Supplement
Quantity 10/Pk.
Yield Each vial for 500mL medium