missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ Skirrow Campylobacter Selective Supplement

Catalog No. OXSR0069H
Change view
Click to view available options
Yield:
Each vial for 2L medium
Each vial for 500mL medium
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Yield
OXSR0069H Each vial for 2L medium
OXSR0069E Each vial for 500mL medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. OXSR0069H Supplier Thermo Scientific™ Supplier No. SR0069H

Oxoid Campylobacter Selective Supplement, Skirrow prepares Skirrow Selective Medium for isolation of Campylobacter spp. at 42°C.

Oxoid Campylobacter Selective Supplement, Skirrow is used for preparation of Skirrow Selective Medium for the isolation of Campylobacter spp. at 42°C.

Add to Blood Agar Base No. 2, Part No. CM0271B or Columbia Blood Agar Base, Part No. CM0331B

Specifications

Description Campylobacter Selective Supplement, Skirrow
Form Freeze-dried
Product Type Selective Supplement
Quantity 10/Pk.
Yield Each vial for 2L medium