missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ Salmonella Selective Supplement

Catalog No. OXSR0194E
Change view
Click to view available options
Quantity:
10/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OXSR0194E 10/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OXSR0194E Supplier Thermo Scientific™ Supplier No. SR0194E

Oxoid Salmonella Selective Supplement is used for the presumptive identification of Salmonella species from clinical and food samples.

Oxoid Salmonella Selective Supplement is used for the presumptive identification of Salmonella species from clinical and food samples.

  • Add to Chromogenic Salmonella Agar Base, Part No. CM1007B or Brilliance™ Salmonella Agar Base, Part No. CM1092B
  • Each vial supplements 500mL of medium

Specifications

Description Salmonella Selective Supplement
Form Freeze-dried
Product Type Selective Supplement
Quantity 10/Pk.
Yield Each vial for 500mL medium
Species Salmonella species