Learn More
Thermo Scientific™ Oxoid™ RPF Supplement
Numéro de catalogue. OXSR0122A
Change view
Click to view available options
Quantity:
10/Pk.
Numéro de catalogue. Quantity
OXSR0122A 10/Pk.
1 options
Numéro de catalogue. OXSR0122A Fournisseur Thermo Scientific™ Code fournisseur SR0122A
Il en reste null
The Oxoid RPF Supplement (Rabbit Plasma Fibrinogen) is used for isolation and enumeration of coagulase positive staphylococci from food.
Oxoid RPF Supplement (Rabbit Plasma Fibrinogen) is used for the isolation and enumeration of coagulase positive staphylococci from food.
  • Use only with Baird-Parker Agar Base, RPF, Part No. CM0961B
  • Each vial supplements 100mL of medium
Spécifications
Description RPF Supplement
Form Freeze-dried
Product Type Microbiology Media
Quantity 10/Pk.