missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ Purified Agar

Catalog No. OXLP0028B
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OXLP0028B 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OXLP0028B Supplier Thermo Scientific™ Supplier No. LP0028B

Oxoid Purified Agar is an exceptionally bright and clear agar recommended for electrophoresis studies but also suitable for many bacteriological purposes.

Oxoid Purified Agar is an exceptionally bright and clear agar recommended for electrophoresis studies but also suitable for many bacteriological purposes.

Specifications

Description Dehydrated; Purified agar
Form Powder
Product Type Agar
Quantity 500 g
Yield For 10.3L medium

For Laboratory Use Only