missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ Perfringens TSC Supplement

Catalog No. OXSR0088E
Change view
Click to view available options
Quantity:
10/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OXSR0088E 10/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OXSR0088E Supplier Thermo Scientific™ Supplier No. SR0088E

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Oxoid Perfringens TSC Supplement (D-cycloserine 200mg/vial) is used for the recovery of Clostridium perfringens.

Oxoid Perfringens TSC Supplement (D-cycloserine 200mg/vial) is used for the recovery of Clostridium perfringens.

  • Add to DCLS Agar, Part No. CM0393B, or Perfringens Agar Base, TSC, Part No. CM0587B
  • Each vial supplements 500mL of medium

Specifications

Description Perfringens TSC Supplement
Form Freeze-dried
Product Type Microbiology Media
Quantity 10/Pk.
Yield Each vial for 500mL medium
Species Perfringens