missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Oxoid™ Perfringens OPSP Selective Supplement A
Description
Oxoid Perfringens OPSP Selective Supplement A is for selective isolation of Clostridium perfringens from food.
- Add to Perfringens Agar Base, OPSP, Part No. CM0543B
- For 500mL of medium
Specifications
Specifications
| Description | Perfringens OPSP Supplement A |
| Form | Freeze-dried |
| Product Type | Selective Supplement |
| Quantity | 10/Pk. |
| Yield | Each vial for 500mL medium |
| Species | Perfringens |