Learn More
Thermo Scientific™ Oxoid™ Peptone P
Catalog No. OXLP0049B
Change view
Click to view available options
Quantity:
500g
Catalog No. Quantity
OXLP0049B 500g
1 options
Catalog No. OXLP0049B Supplier Thermo Scientific™ Supplier No. LP0049B
Thermo Scientific™ Oxoid Peptone P is a peptic digest which conforms to the USP specification of peptic digests of animal tissue.
Thermo Scientific™ Oxoid Peptone P is a peptic digest which conforms to the USP specification of peptic digests of animal tissue.
  • The high sulphur content makes it ideal for use in culture media
Specifications
Description Dehydrated; Peptone P
Form Powder
Product Type Peptone
Quantity 500g