missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ Peptone Bacteriological Neutralized

Numéro de catalogue. OXLP0034K
Modifier l'affichage
Click to view available options
Quantity:
500 g
25 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
OXLP0034K 25 kg
OXLP0034B 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. OXLP0034K Fournisseur Thermo Scientific™ Code fournisseur LP0034K
Il en reste null

Oxoid Peptone Bacteriological Neutralized is the neutralized form of Peptone Bacteriological.

Oxoid Peptone Bacteriological Neutralized is the neutralized form of Peptone Bacteriological.

Spécifications

Description Peptone Bacteriological Neutralized
Form Powder
Product Type Peptone
Quantity 25 kg