missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ Peptone Bacteriological

Catalog No. OXLP0037B
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OXLP0037B 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OXLP0037B Supplier Thermo Scientific™ Supplier No. LP0137B

The Oxoid Peptone Bacteriological is an all-purpose peptone specially prepared for use in fermentation applications.

Thermo Scientific™ Oxoid Peptone Bacteriological and its neutralized form are very nutritious, all-purpose peptones prepared by the enzymatic digestion of selected animal protein sources. They are specially prepared to be compatible with other refined culture media or fermentation ingredients.

The combination of pancreatin and papain enzyme systems ensures that these bacteriological peptones contain a wide spectrum of polypeptides, reflected in their broad molecular profiles. 

The neutralized form evolved from the original to meet those occasions when a slightly higher pH is required. Both, when reconstituted, give a solution free of haze, cloudiness, or precipitation.

A nutritious, all-purpose peptone specially prepared for use with other refined culture media ingredients

Specifications

Content And Storage Dehydrated Powder, Ambient & Dry Storage
Form Dehydrated Powder
Product Type Peptone
For Use With (Application) Culture media or fermentation processes
Quantity 500 g
Protein Source Beef

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.