Learn More
Thermo Scientific™ Oxoid™ PALCAM Selective Supplement
Catalog No. OXSR0150E
Change view
Click to view available options
Description:
PALCAM Selective Supplement
Catalog No. Description
OXSR0150E PALCAM Selective Supplement
1 options
Catalog No. OXSR0150E Supplier Thermo Scientific™ Supplier No. SR0150E
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Oxoid PALCAM Selective Supplement is used for the detection of Listeria monocytogenes.
Oxoid PALCAM Selective Supplement is used for the detection of Listeria monocytogenes.
Add to PALCAM Agar Base, Part No. CM0877B
Specifications
Description PALCAM Selective Supplement
Form Freeze-dried
Product Type Selective Supplement
Quantity 10/Pk.
Yield Each vial for 500mL medium