Learn More
Thermo Scientific™ Oxoid™ ORSAB Selective Supplement
Description
Oxoid ORSAB Selective Supplement (Oxacillin Resistance Screening Agar Base Selective Supplement) is added to Oxacillin Resistance Screening Agar Base, Part No. CM1008B.
- Add to Oxacillin Resistance Screening Agar Base, Part No. CM1008B
- Each vial supplements 500mL of medium
Specifications
Specifications
| Description | ORSAB Selective Supplement |
| Form | Freeze-dried |
| Product Type | Selective Supplement |
| Quantity | 10/Pk. |
| Yield | Each vial for 500mL medium |