Learn More
Thermo Scientific™ Oxoid™ OGYE Selective Supplement
Catalog No. OXSR0073A
Change view
Click to view available options
Quantity:
10/Pk.
Catalog No. Quantity
OXSR0073A 10/Pk.
1 options
Catalog No. OXSR0073A Supplier Thermo Scientific™ Supplier No. SR0073A
Oxoid OGYE Selective Supplement (Oxytetracycline Glucose Yeast Extract) is added to Oxytetracycline Glucose Yeast Extract Agar Base, Part No. CM0545B.
Oxoid OGYE Selective Supplement (Oxytetracycline Glucose Yeast Extract) is added to Oxytetracycline Glucose Yeast Extract Agar Base, Part No. CM0545B.
  • Add to Oxytetracycline Glucose Yeast Extract Agar Base, Part No. CM0545B
  • Each vial supplements 500mL of medium
Specifications
Description OGYE Selective Supplement
Form Freeze-dried
Product Type Selective Supplement
Quantity 10/Pk.
Yield Each vial for 500mL medium