Learn More
Thermo Scientific™ Oxoid™ Novobiocin Selective Supplement
Description
Oxoid Novobiocin Selective Supplement (10mg/vial) is added to EC Broth, Part No. CM0990B; Muller-Kauffmann Tetrathionate-Novobiocin Broth Base, Part No. CM0343B; Brilliant Green Agar, Part No. CM0263B; MRSV, Part No. CM0910B; Modified Tryptone Soya Broth, Part No. CM0989B; and Lysine Iron Agar, Modified, Part No. CM0886B.
- Each vial supplements 500mL of medium
Specifications
Specifications
| Description | Novobiocin Selective Supplement |
| Form | Freeze-dried |
| Product Type | Selective Supplement |
| Quantity | 10 x 10 mg |
| Yield | Yield will be dependent upon formulation; follow the guidelines for your specific product. |