Learn More
Thermo Scientific™ Oxoid™ Neomycin Selective Supplement
Description
Oxoid Neomycin Selective Supplement (150mg/vial) allows the growth of clostridia and other anaerobes.
- Add to Blood Agar, Wilkins-Chalgren Anaerobe Agar, Part No. CM0619B, or Columbia Blood Agar, Part No. CM0331B
- Each vial supplements 2L of medium
Specifications
Specifications
| Description | Neomycin Selective Supplement |
| Form | Freeze-dried |
| Product Type | Selective Supplement |
| Quantity | 10 x 150 mg |
| Yield | Each vial for 2L medium |