Learn More
Thermo Scientific™ Oxoid™ MRSV Selective Supplement
Catalog No. OXSR0161E
Change view
Click to view available options
Quantity:
10/Pk.
Catalog No. Quantity
OXSR0161E 10/Pk.
1 options
Catalog No. OXSR0161E Supplier Thermo Scientific™ Supplier No. SR0161E
Oxoid MRSV Selective Supplement is used for the detection of Salmonella spp. from food and environmental samples.
Oxoid MRSV Selective Supplement is used for the detection of Salmonella spp. from food and environmental samples.
  • Add to MSRV Medium Base, Part No. CM0910B and Lysine Iron Agar, Modified (MLIA), Part No. CM0886B
  • Each vial supplements 500mL of medium
Specifications
Description MSRV Selective Supplement
Form Freeze-dried
Product Type Selective Supplement
Quantity 10/Pk.
Yield Each vial for 500mL medium