missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ Modified Preston Campylobacter Selective Supplement

Catalog No. OXSR0204E
Change view
Click to view available options
Quantity:
10/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OXSR0204E 10/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OXSR0204E Supplier Thermo Scientific™ Supplier No. SR0204E

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Used for the isolation of Campylobacter jejuni and Campylobacter coli from human, animal, avian, and environmental samples.

Oxoid Modified Preston Campylobacter Selective Supplement is for the isolation of Campylobacter jejuni and Campylobacter coli from human, animal, avian, and environmental samples.

  • Alternative supplement containing amphotericin B as a replacement for cycloheximide
  • Add to Campylobacter Agar Base, Part No. CM0689B or Nutrient Broth No. 2, Part No. CM0067B
  • Each vial supplements 500mL of medium

Specifications

Description Modified Preston Campylobacter Selective Supplement
Form Freeze-dried
Product Type Selective Supplement
Quantity 10/Pk.
Yield Each vial for 500mL medium
Species Campylobacter