missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ Modified Karmali Selective Supplement

Catalog No. OXSR0205E
Change view
Click to view available options
Quantity:
10/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OXSR0205E 10/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OXSR0205E Supplier Thermo Scientific™ Supplier No. SR0205E

A blood-free selective medium for the isolation of Campylobacter jejuni and Campylobacter coli when incubated at 42° C.

Oxoid Modified Karmali Selective Supplement is for a blood-free selective medium for the isolation of Campylobacter jejuni and Campylobacter coli when incubated at 42° C.

  • Alternative supplement containing amphotericin B as a replacement for cycloheximide
  • Add to Campylobacter Agar Base (Karmali), Part No. CM0935B
  • Each vial supplements 500mL of medium

Specifications

Description Modified Karmali Selective Supplement
Form Freeze-dried
Product Type Selective Supplement
Quantity 10/Pk.
Yield Each vial for 500mL medium
Species Karmali