missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ Modified Brucella Selective Supplement

Catalog No. OXSR0209E
Change view
Click to view available options
Quantity:
10/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OXSR0209E 10/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OXSR0209E Supplier Thermo Scientific™ Supplier No. SR0209E

Oxoid Modified Brucella Selective Supplement is for the cultivation and isolation of Brucella spp.

Oxoid Modified Brucella Selective Supplement is for the cultivation and isolation of Brucella spp.

  • Alternative supplement containing natamycin as a replacement for cycloheximide
  • Add to Blood Agar Base No. 2, Part No. CM0271B or Brucella Medium Base, Part No. CM0169B
  • Each vial supplements 500mL of medium

Specifications

Description Modified Brucella Selective Supplement
Form Freeze-dried
Product Type Selective Supplement
Quantity 10/Pk.
Yield Each vial for 500mL medium
Species Brucella species