missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Oxoid™ Modified Bolton Broth Selective Supplement
Description
Oxoid Modified Bolton Broth Selective Supplement is for the selective pre-enrichment of Campylobacter spp. in food samples.
- Alternative supplement containing amphotericin B as a replacement for cycloheximide
- Add to Bolton Broth, Part No. CM0983B
- Each vial supplements 500mL of medium
Specifications
Specifications
| Description | Modified Bolton Broth Selective Supplement |
| Form | Freeze-dried |
| Product Type | Selective Supplement |
| Quantity | 10/Pk. |
| Yield | Each vial for 500mL medium |
| Species | Bolton |