missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ Membrane Clostridium Perfringens Selective Supplement

Catalog No. OXSR0188E
Change view
Click to view available options
Quantity:
10/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OXSR0188E 10/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OXSR0188E Supplier Thermo Scientific™ Supplier No. SR0188E

A chromogenic medium for rapid and presumptive identification of Clostridium perfringens from water samples.

Oxoid Membrane Clostridium Perfringens Selective Supplement (m-CP) is a chromogenic medium for rapid and presumptive identification of Clostridium perfringens from water samples.

  • Add to Membrane Clostridium Perfringens (m-CP) Agar Base, Part No. CM0992B
  • Each vial of SR0188E supplements 500mL of medium

Specifications

Description Membrane Clostridium Perfringens Selective Supplement
Form Freeze-dried
Product Type Selective Supplement
Quantity 10/Pk.
Yield Each vial for 500mL medium
Species Perfringens