Learn More
Thermo Scientific™ Oxoid™ Malt Extract
Oxoid Malt Extract is recommended for use in media for yeast and mold growth.
$271.26 - $271.26
Specifications
Form Powder
Product Type Malt Extract
View More Specs
Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
OXLP0039B
View Documents
Thermo Scientific™
LP0039B
500 g
Each for $271.26
Only null left
Description
Description
Oxoid Malt Extract is recommended for use in media for yeast and mold growth.
An extract of sprouted grain with conserved nitrogen and carbohydrate constituents
Specifications
Specifications
Powder
Malt Extract
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'