Learn More
Thermo Scientific™ Oxoid™ Kanamycin Sulphate Supplement
Description
Oxoid Kanamycin Sulphate Supplement (10mg/vial) is added to Kanamycin Esculin Azide Agar Base, Part No. CM0591B.
Add to Kanamycin Esculin Azide Agar Base, Part No. CM0591B
Specifications
Specifications
| Description | Kanamycin Sulphate Supplement |
| Form | Freeze-dried |
| Product Type | Microbiology Media |
| Quantity | 10/Pk. |
| Yield | Each vial for 500mL medium |