missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ Half Fraser Supplement

Catalog No. OXSR0166G
Change view
Click to view available options
Quantity:
10/Pk.
For 2.25L of base
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OXSR0166G For 2.25L of base
OXSR0166E 10/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. OXSR0166G Supplier Thermo Scientific™ Supplier No. SR0166G

The Oxoid Half Fraser Supplement is used for the enrichment of Listeria species.

Oxoid Half Fraser Supplement is used for the enrichment of Listeria species.

  • Add to Fraser Broth Base, Part No. CM0895B

Specifications

Description Half Fraser Supplement
Form Freeze-dried
Product Type Microbiology Media
Quantity For 2.25L of base
Species Listeria