Learn More
Thermo Scientific™ Oxoid™ Dextrose Bacteriological
Catalog No. OXLP0071B
Change view
Click to view available options
Quantity:
500 g
Catalog No. Quantity
OXLP0071B 500 g
1 options
Catalog No. OXLP0071B Supplier Thermo Scientific™ Supplier No. LP0071B
Oxoid Dextrose Bacteriological is a sugar suitable for inclusion in bacteriological culture media.
Oxoid Dextrose Bacteriological is a sugar suitable for inclusion in bacteriological culture media.
Specifications
Description Dehydrated; Dextrose bacteriological
Form Powder
Product Type Media Supplement
Quantity 500 g