missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ Cycloheximide Solution (0.1%)

Catalog No. OXSR0222C
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OXSR0222C 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OXSR0222C Supplier Thermo Scientific™ Supplier No. SR0222C

Oxoid Cycloheximide Solution (0.1%) (100mg/vial) is used for the suppression of fungi.

Oxoid Cycloheximide Solution (0.1%) (100mg/vial) is used for the suppression of fungi.

Add to Bacillus Cereus Selective Agar Base, Part No. CM0617B; Raka-Ray Agar Base, Part No. CM0777B; and Universal Beer Agar, Part No. CM0651B

Specifications

Description Cycloheximide Solution (0.1%)
Form Liquid
Product Type Solution
Quantity 100 mL
Yield Yield will be dependent upon formulation; follow the guidelines for your specific product.