missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ Chloramphenicol Selective Supplement

Catalog No. p-4522523
Change view
Click to view available options
Yield:
Each vial for 2L medium
Each vial for 500mL medium
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Yield
OXSR0078E Each vial for 500mL medium
OXSR0078H Each vial for 2L medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. OXSR0078E Supplier Thermo Scientific™ Supplier No. SR0078E
Only null left

Oxoid Chloramphenicol Selective Supplement assists in the isolation of yeast and molds associated with spoilage.

For the isolation of yeast and molds associated with spoilage, add Oxoid Chloramphenicol Selective Supplement to DRBC Agar Base, Part No. CM0727B; for the enumeration of yeast and molds in foods, add to Rose Bengal Chloramphenicol Agar Base, Part No. CM0549B.

For the isolation of yeast and molds associated with spoilage, add to DRBC Agar Base, Part No. CM0727B; for the enumeration of yeast and molds in foods, add to Rose Bengal Chloramphenicol Agar Base, Part No. CM0549B.

Specifications

Description Chloramphenicol Selective Supplement
Form Freeze-dried
Product Type Selective Supplement
Quantity 10/Pk.
Yield Each vial for 500mL medium