Learn More
Thermo Scientific™ Oxoid™ Cefixime Supplement
Catalog No. OXSR0191E
Change view
Click to view available options
Quantity:
10/Pk.
Catalog No. Quantity
OXSR0191E 10/Pk.
1 options
Catalog No. OXSR0191E Supplier Thermo Scientific™ Supplier No. SR0191E
Only null left
Oxoid Cefixime Supplement is used for the isolation of Escherichia coli O157.
Oxoid Cefixime Supplement is used for the isolation of Escherichia coli O157.
  • Add to to Cefixime Rhamnose Sorbitol MacConkey Agar Base, Part No. CM1005B
  • Each vial supplements 500mL of medium
Specifications
Description Cefixime Supplement
Form Freeze-dried
Product Type Microbiology Media
Quantity 10/Pk.