missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ Casein Hydrolysate

Numéro de catalogue. OXLP0041B
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
OXLP0041B 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. OXLP0041B Fournisseur Thermo Scientific™ Code fournisseur LP0041B
Il en reste null

Oxoid Casein Hydrolysate (Acid) is a purified hydrolysate high in nitrogenous material ideal for culture media or as an amino acid mixture.

Thermo Scientific™ Oxoid Casein Hydrolysate (Acid) is a purified hydrolysate which provides the necessary nitrogenous material for culture media or can be used in fermentation when amino acid mixtures are specified.

Thermo Scientific™ Oxoid Casein Hydrolysate (Acid) is a purified hydrolysate which provides the necessary nitrogenous material for culture media or can be used in fermentation when amino acid mixtures are specified.

With over 100 years of culture media manufacturing experience, from raw materials to final product, we offer the expertise, quality assurance, and scale to ensure successful microbial vaccine production.

Our Thermo Scientific Oxoid peptones are available in both animal-origin and animal-origin-free options, offering a range of AN/TN ratios to suit the specific needs of your process.

Supports quality media design:

  • Balanced nutrient composition
  • Batch-to-batch consistency, available in sizes from 500 g up to 25 kg and in single batch sizes up to 2.5 T
  • Highly soluble
  • BSE and mycoplasma free

Multi-method suitability:

  • Batch culture
  • Fed-batch culture
  • Continuous culture
  • Cell culture

Spécifications

Content And Storage Dehydrated Powder, Ambient & Dry Storage
Description Dehydrated; Casein hydrolysate (Acid)
Form Dehydrated Powder
Product Type Peptone
For Use With (Application) Culture media or fermentation processes
Quantity 500 g
Protein Source Casein

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.