missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Oxoid™ Campylobacter Growth Supplement
Description
Oxoid Campylobacter Growth Supplement is a liquid supplement for the enhanced growth and aerotolerance of Campylobacter species.
- For the preparation of Blaser-Wang Medium, Bolton Selective Enrichment Broth, Butzler Selective Medium, and Preston Selective Medium
- Each vial supplements 500mL of medium
Specifications
Specifications
| Description | Campylobacter Growth Supplement |
| Form | Freeze-dried |
| Product Type | Microbiology Media |
| Quantity | 10/Pk. |
| Yield | Each vial for 500mL medium |
| Species | Campylobacter |