missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ C-T Supplement

Catalog No. OXSR0172H
Change view
Click to view available options
Yield:
Each vial for 2L medium
Each vial for 500mL medium
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Yield
OXSR0172H Each vial for 2L medium
OXSR0172E Each vial for 500mL medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. OXSR0172H Supplier Thermo Scientific™ Supplier No. SR0172H

Oxoid C-T Supplement is a chromogenic medium for the identification of Escherichia coli O157:H7.

Oxoid C-T Supplement is a chromogenic medium for the identification of Escherichia coli O157:H7.

Add to Sorbitol MacConkey Agar, Part No. CM0813B or to Sorbitol MacConkey Agar with BCIG, Part No. CM0981B.

Ordering Information:

Each vial supplements 2L of medium.

Specifications

Description C-T Supplement
Form Freeze-dried
Product Type Microbiology Media
Quantity 10/Pk.
Yield Each vial for 2L medium