missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ Butzler Campylobacter Selective Supplement

Catalog No. OXSR0085E
Change view
Click to view available options
Quantity:
10/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OXSR0085E 10/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OXSR0085E Supplier Thermo Scientific™ Supplier No. SR0085E

The Oxoid Campylobacter Selective Supplement, Butzler is used for the isolation of Campylobacter spp. at 37°C.

Oxoid Campylobacter Selective Supplement, Butzler is used for the isolation of Campylobacter spp. at 37°C.

  • Add to Columbia Blood Agar Base, Part No. CM0331B
  • Each vial supplements 500mL of medium

Specifications

Description Campylobacter Selective Supplement, Butzler
Form Freeze-dried
Product Type Selective Supplement
Quantity 10/Pk.
Yield Each vial for 500mL medium
Species Campylobacter