missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ Burkholderia Cepacia Selective Supplement

Catalog No. OXSR0189E
Change view
Click to view available options
Quantity:
10/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OXSR0189E 10/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OXSR0189E Supplier Thermo Scientific™ Supplier No. SR0189E

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Oxoid Burkholderia Cepacia Selective Supplement is used for the isolation of Burkholderia cepacia.

Oxoid Burkholderia Cepacia Selective Supplement is used for the isolation of Burkholderia cepacia.

  • Add to Burkholderia Cepacia Agar Base, Part No. CM0995B
  • Each vial supplements 500mL of medium

Specifications

Description Burkholderia Cepacia Selective Supplement
Form Freeze-dried
Product Type Selective Supplement
Quantity 10/Pk.
Yield Each vial for 500mL medium
Species Burkholderia cepacia