Learn More
Thermo Scientific™ Oxoid™ Broad Spectrum Mixture Beta Lactamase
Catalog No. OXSR0113E
Change view
Click to view available options
Quantity:
10/Pk.
Catalog No. Quantity
OXSR0113E 10/Pk.
1 options
Catalog No. OXSR0113E Supplier Thermo Scientific™ Supplier No. SR0113E
Mixture of beta-lactamase enzymes from Bacillus cereus for the inactivation of beta-lactam antibiotic preparations present in specimens.
Oxoid Beta Lactamase, Broad Spectrum Mixture is a mixture of beta-lactamase enzymes from Bacillus cereus for the inactivation of beta-lactam antibiotic preparations present in specimens.
Specifications
Description Beta Lactamase, Broad Spectrum Mixture
Form Freeze-dried
Product Type Beta Lactamase Mixture
Quantity 10/Pk.