missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Brilliance Bacillus Cereus Selective Supplement

Catalog No. OXSR0230E
Change view
Click to view available options
Quantity:
10/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OXSR0230E 10/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OXSR0230E Supplier Thermo Scientific™ Supplier No. SR0230E

Brilliance is a chromogenic medium for the isolation and differentiation of Bacillus cereus in food samples.

Add to Brilliance Bacillus Cereus Agar Base, Part No. CM1036A and CM1036B. Each vial supplements 500mL of medium.

Specifications

Description Brilliance Bacillus Cereus Selective Supplement
Form Freeze-dried
Product Type Selective Supplement
Quantity 10/Pk.
Yield Each vial for 500mL medium
Species Bacillus cereus