missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ Bordetella Selective Supplement

Catalog No. OXSR0082E
Change view
Click to view available options
Quantity:
10/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OXSR0082E 10/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OXSR0082E Supplier Thermo Scientific™ Supplier No. SR0082E

Oxoid Bordetella Selective Supplement (Cephalexin, 20mg/vial) isolates Bordetella pertussis and B. parapertussis.

Oxoid Bordetella Selective Supplement (Cephalexin, 20mg/vial) is used for the selective isolation of Bordetella pertussis and B. parapertussis.

Add to to Charcoal Agar Base, Part No. CM0119B. Each vial supplements 500mL of medium.

Specifications

Description Bordetella Selective Supplement
Form Freeze-dried
Product Type Selective Supplement
Quantity 10/Pk.
Yield Each vial for 500mL medium
Species Bordetella species