missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ Bolton Broth Selective Supplement

Catalog No. OXSR0183E
Change view
Click to view available options
Quantity:
10/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OXSR0183E 10/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OXSR0183E Supplier Thermo Scientific™ Supplier No. SR0183E

Oxoid Bolton Broth Selective Supplement is used for the selective pre-enrichment of Campylobacter spp. in food samples.

Oxoid Bolton Broth Selective Supplement is used for the selective pre-enrichment of Campylobacter spp. in food samples.

Add to to Bolton Broth, Part No. CM0983B. Each vial supplements 500mL of medium.

Specifications

Description Bolton Broth Selective Supplement
Form Freeze-dried
Product Type Selective Supplement
Quantity 10/Pk.
Yield Each vial for 500mL medium
Species Bolton