Learn More
Thermo Scientific™ Oxoid™ Bile Salts No. 3
The Oxoid Bile Salts No. 3 is a specifically refined fraction of bile acid salts.
$943.06 - $943.06
Specifications
Form Powder
Product Type Bile Salts
View More Specs
Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
OXLP0056J
View Documents
Thermo Scientific™
LP0056J
250 g N/A
Only null left
Description
Description
Oxoid Bile Salts No. 3 is a specifically refined fraction of bile acid salts.
Normal working concentration 0.15% w/v. Each vial supplements 500 mL of medium.
Specifications
Specifications
Powder
Bile Salts
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'