Learn More
Thermo Scientific™ Oxoid™ Bile Salts
Oxoid Bile Salts is a standardized ox-bile extract used as a selective inhibitory agent in culture media.
$541.06 - $541.06
Specifications
Form Powder
Product Type Bile Salts
View More Specs
Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
OXLP0055J
View Documents
Thermo Scientific™
LP0055J
250 g
Each for $541.06
Only null left
Description
Description
Oxoid Bile Salts is a standardized ox-bile extract used as a selective inhibitory agent in culture media.
Normal working concentration 0.5% w/v. Each vial supplements 500 mL of medium.
Specifications
Specifications
Powder
Bile Salts
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'