missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Oxoid™ Bacillus Cereus Selective Supplement

Catalog No. OXSR0099E
Change view
Click to view available options
Quantity:
10 x 50,000 IU
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OXSR0099E 10 x 50,000 IU
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OXSR0099E Supplier Thermo Scientific™ Supplier No. SR0099E

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Oxoid Bacillus Cereus Selective Supplement (Polymyxin B Supplement) is supplied 50,000 IU/Vial for inhibition of Gram-negative bacteria.

Oxoid Bacillus Cereus Selective Supplement (Polymyxin B Supplement) is supplied 50,000 IU/Vial for inhibition of Gram-negative bacteria.

  • May be added to Bacillus cereus Selective Agar Base, Part No. CM0617B and MYP Agar Base, Part No. CM0929B
  • Each vial supplements 500mL of medium

Specifications

Description Bacillus Cereus Selective Supplement
Form Freeze-dried
Product Type Selective Supplement
Quantity 10 x 50,000 IU
Yield Each vial for 500mL medium
Species Bacillus cereus