missing translation for 'onlineSavingsMsg'
Learn More

Oxindole, 97+%

Catalog No. aaa1876414
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1876414 25 g
AAA1876406 5 g
AAA1876422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1876414 Supplier Thermo Scientific Chemicals Supplier No. A1876414
Only null left

CAS: 59-48-3 | C8H7NO | 133.15 g/mol

Oxindole is considered as indole analogue, which shows pharmacological activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Oxindole is considered as indole analogue, which shows pharmacological activity.

Solubility
Soluble in methanol, ethanol, dimethyl sulfoxide and ether. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 59-48-3
Molecular Formula C8H7NO
Molecular Weight (g/mol) 133.15
MDL Number MFCD00005711
InChI Key JYGFTBXVXVMTGB-UHFFFAOYSA-N
Synonym oxindole, indolin-2-one, 2-oxindole, 2-indolinone, 1,3-dihydro-2h-indol-2-one, oxindol, 2-oxoindoline, 2-oxindoline, indol-2 3h-one, 2h-indol-2-one, 1,3-dihydro
PubChem CID 321710
ChEBI CHEBI:31697
IUPAC Name 1,3-dihydroindol-2-one
SMILES C1C2=CC=CC=C2NC1=O

Specifications

Melting Point 124°C to 128°C
Boiling Point 194°C to 196°C (17 mmHg)
Quantity 25 g
Beilstein 114692
Solubility Information Soluble in methanol,ethanol,dimethyl sulfoxide and ether. Insoluble in water.
Formula Weight 133.15
Percent Purity ≥97%
Chemical Name or Material Oxindole
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 200-429-5
RTECSNumber NM2080500
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only