missing translation for 'onlineSavingsMsg'
Learn More

[(Oxido)phenyl(trifluoromethyl)-lambda4-sulfanylidene]dimethylammonium Tetrafluoroborate 98.0+%, TCI America™

Catalog No. p-7134298
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O0367200MG 200 mg
O03671G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O0367200MG Supplier TCI America Supplier No. O0367200MG

Chemical Identifiers

CAS 1046786-08-6
Molecular Formula C9H11BF7NOS
Synonym Shibata Reagent I

Specifications

Color White
Quantity 200 mg
UN Number 1759
Formula Weight 325.05
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material [(Oxido)phenyl(trifluoromethyl)-lambda4-sulfanylidene]dimethylammonium Tetrafluoroborate