missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Oxidized Low-density Lipoprotein (OxLDL) and DiI-OxLDL

Catalog No. L34357
Encompass
Change view
Click to view available options
Conjugate:
Dil-OxLDL
Ox-LDL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Conjugate
L34357 Ox-LDL
L34358 Dil-OxLDL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. L34357 Supplier Invitrogen™ Supplier No. L34357
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Study lipid transport and atherosclerosis with oxidized LDL (OxLDL) and the fluorescently labeled DiI-OxLDL. Oxidized forms of human low-density lipoprotein (LDL) are used to examine scavenger receptor-mediated endocytosis of OxLDL by macrophages and endothelial cells.

Oxidized LDL (OxLDL) and the fluorescently labeled DiI-OxLDL are oxidized forms of the human low-density lipoprotein (LDL). LDL is an important lipid-protein complex that is involved in lipid transport and contributes to atherosclerosis. These unlabeled and fluorescent OxLDL probes are used to study scavenger receptor-mediated endocytosis by macrophages and endothelial cells, and the formation of macrophage-derived foam cells that occurs in early atherogenesis.

Human low-density lipoprotein (LDL) is one of the key lipid-protein complexes in blood and is a crucial component of metabolism responsible for the transport of lipids throughout the body. LDL is composed of cholesterol, cholesterol esters, triglycerides, phospholipids, and a single Apo B-100 protein. LDLs deliver fatty acids and cholesterol to peripheral and liver cells through receptor-mediated endocytosis. High levels of LDL cholesterol are considered a risk factor for atherosclerosis and cardiovascular disease, while high levels of high-density lipoproteins (HDLs) may reduce the risk.

Oxidation of LDL is a natural process within the body caused by free radicals, however excess oxidized LDL can be harmful. Alterations in lipid metabolism such as oxidation of LDL can lead to inflammation in arteries and atherosclerosis. Atherosclerosis is a type of arteriosclerosis in which arterial wall thickening is due to the buildup of fatty plaques, composed of cholesterol and other lipids, that can inhibit and block blood flow in vessels. Endothelial cells and macrophages are recruited to remove oxidized LDL (OxLDL), which initiates inflammatory and immunogenic responses. Unlabeled and fluorescent OxLDL probes are important tools in the study of scavenger receptor-mediated endocytosis by macrophages and endothelial cells, as well as the formation of macrophage-derived foam cells, a hallmark of early atherogenesis.

The OxLDL probe is generated by oxidizing the surface lipids of native unmodified LDL by incubating with copper sulfate. Oxidation is monitored during the incubation by measuring the optical density at 234 and terminated at approximately the halfway point during the lipid peroxidation phase to a level of approximately 25–35 nmol/mg protein in TBAR (thiobarbituric acid–reactive) values. This controlled oxidation procedure is used to ensure that lipids on the surface of the LDL are oxidized, with limited oxidation of the surface apolipoprotein, allowing the OxLDL to induce a physiologically relevant inflammatory response from cells.

Features of OxLDL:
• Oxidized by copper-mediated process
• Each lot tested for optimal level of oxidation and functionally tested with Bovine Pulmonary Artery Epithelium (BPAE) cells for recognition by scavenger receptors
• Can be used as a negative control to block cell surface receptors or to stimulate inflammatory/immunogenic responses and in applications such as ELISA and Western blot

Oxidized LDL is also complexed with the red-orange fluorescent DiI dye (DiI-OxLDL) to enable fluorescence detection of oxidized LDL uptake. DiI is a highly fluorescent lipophilic dye that diffuses into the hydrophobic portion of the LDL complex and does not affect the LDL-specific binding of the apolipoprotein. The DiI-OxLDL degree of labeling has been optimized for superior sensitivity in various fluorescence applications such as fluorescence microscopy, flow cytometry, cell sorting, high-throughput analysis, and high-content analysis.

Features of DiI-OxLDL:
• Detectable with RFP/TRITC filter set (absorption peak: ∼554 nm, emission peak: ∼571 nm)
• Fixable and multiplexable with green, deep red, violet, and blue emission dyes, such as GFP, DAPI, Hoechst, HCS LipidTOX, and LysoTracker Deep Red
• Each lot functionally tested with Bovine Pulmonary Artery Epithelium (BPAE) cells for recognition by scavenger receptors
• Optimized oxidization and degree of labeling for superior sensitivity

Specifications

Content And Storage 1 x 200 μL OxLDL, 2.5 mg/mL
Store in refrigerator (2°C to 8°C) and protect from light.
For Use With (Application) Cell Viability, Cell Proliferation and Function
Form Solution
Product Line Molecular Probes
Quantity 200 μL
Shipping Condition Wet Ice
Species Human
Protein Family LDL
Conjugate Ox-LDL
If the Low Density Lipoprotein (LDL) from Human Plasma was accidentally frozen, can I still use it?

We recommend storing Low Density Lipoprotein (LDL) from Human Plasma at 2-8 degrees C. We do not recommend using frozen product.

Can I use Low Density Lipoprotein (LDL) from Human Plasma on other species?

We have not tested this.