missing translation for 'onlineSavingsMsg'
Learn More

Oxibendazole 98.0+%, TCI America™

Catalog No. p-7134365
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O0499100MG 100 mg
O0499500MG 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O0499100MG Supplier TCI America Supplier No. O0499100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 20559-55-1
Molecular Formula C12H15N3O3
Molecular Weight (g/mol) 249.27
MDL Number MFCD00133728
InChI Key RAOCRURYZCVHMG-UHFFFAOYSA-N
Synonym Methyl (5-Propoxy-1H-benzo[d]imidazol-2-yl)carbamate, (5-Propoxy-1H-benzimidazol-2-yl)carbamic Acid Methyl Ester
PubChem CID 4622
IUPAC Name methyl N-(6-propoxy-1H-benzimidazol-2-yl)carbamate
SMILES CCCOC1=CC2=C(C=C1)N=C(N2)NC(=O)OC

Specifications

Melting Point 230°C
Quantity 100 mg
Formula Weight 249.27
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Oxibendazole
TSCA No
Recommended Storage Refrigerator