missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ OXGR1 Recombinant Rabbit Monoclonal Antibody (23GB4575)

Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA553772
Description
OXGR1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
G protein-coupled receptors (GPCRs, or GPRs) contain 7 transmembrane domains and transduce extracellular signals through heterotrimeric G proteins.Chemical Identifiers
| OXGR1 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| OXGR1 | |
| 2-oxoglutarate receptor 1; aKGR; Alpha-ketoglutarate receptor 1; Cysltr3; G protein-coupled receptor 80; G protein-coupled receptor 99; Gm299; Gpr80; Gpr99; G-protein coupled receptor 80; G-protein coupled receptor 99; MGC119206; MGC119207; MGC119208; Oxgr1; oxoglutarate (alpha-ketoglutarate) receptor 1; oxoglutarate receptor 1; P2RY15; P2Y purinoceptor 15; P2Y15; P2Y-like GPCR; P2Y-like nucleotide receptor; seven transmembrane helix receptor | |
| OXGR1 | |
| Primary | |
| Affinity Chromatography |
| 23GB4575 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q6IYF8, Q6Y1R5, Q96P68 | |
| 239283, 27199, 290493 | |
| A synthesized peptide derived from human OXGR1/GPR99 (181-215AA). | |
| Antibody |
Specifications
| OXGR1 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| OXGR1 | |
| 2-oxoglutarate receptor 1; aKGR; Alpha-ketoglutarate receptor 1; Cysltr3; G protein-coupled receptor 80; G protein-coupled receptor 99; Gm299; Gpr80; Gpr99; G-protein coupled receptor 80; G-protein coupled receptor 99; MGC119206; MGC119207; MGC119208; Oxgr1; oxoglutarate (alpha-ketoglutarate) receptor 1; oxoglutarate receptor 1; P2RY15; P2Y purinoceptor 15; P2Y15; P2Y-like GPCR; P2Y-like nucleotide receptor; seven transmembrane helix receptor | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 239283, 27199, 290493 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB4575 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q6IYF8, Q6Y1R5, Q96P68 | |
| OXGR1 | |
| A synthesized peptide derived from human OXGR1/GPR99 (181-215AA). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |