missing translation for 'onlineSavingsMsg'
Learn More

Oxfendazole 98.0+%, TCI America™

Catalog No. O03911G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O03911G 1 g
O03915G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O03911G Supplier TCI America Supplier No. O03911G

Chemical Identifiers

CAS 53716-50-0
Molecular Formula C15H13N3O3S
Molecular Weight (g/mol) 315.347
MDL Number MFCD00801063
InChI Key BEZZFPOZAYTVHN-UHFFFAOYSA-N
Synonym Methyl [5-(Phenylsulfinyl)benzimidazol-2-yl]carbamate, [5-(Phenylsulfinyl)benzimidazol-2-yl]carbamic Acid Methyl Ester
PubChem CID 40854
ChEBI CHEBI:35812
IUPAC Name methyl N-[6-(benzenesulfinyl)-1H-benzimidazol-2-yl]carbamate
SMILES COC(=O)NC1=NC2=C(N1)C=C(C=C2)S(=O)C3=CC=CC=C3

Specifications

Quantity 1 g
Formula Weight 315.35
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Oxfendazole
RTECSNumber FD0808000
TSCA No