Learn More
Oxetane-3-carboxylic acid, 95%
CAS: 114012-41-8 | C4H6O3 | 102.089 g/mol
Supplier: Thermo Scientific Chemicals H57578ME
Description
Oxetane-3-carboxylic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsOxetane-3-carboxylic acid is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.
Solubility
Slightly soluble in water.
Notes
Store in a cool place. Moisture sensitive. . Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Specifications
| Oxetane-3-carboxylic acid | |
| 1.374 | |
| 115°C (239°F) | |
| MFCD08669505 | |
| Air sensitive | |
| Slightly soluble in water. | |
| C1C(CO1)C(=O)O | |
| 102.089 | |
| 102.09 |
| 114012-41-8 | |
| 244°C | |
| C4H6O3 | |
| 500 mg | |
| 3-oxetanecarboxylic acid, 3-oxetanecarboxylicacid, acmc-20ahnr, oxetane-3-carboxylc acd, oxetane-3-carboxylicacid | |
| UWOTZNQZPLAURK-UHFFFAOYSA-N | |
| oxetane-3-carboxylic acid | |
| 19847174 | |
| 95% |
Chemical Identifiers
| 114012-41-8 | |
| 102.089 | |
| UWOTZNQZPLAURK-UHFFFAOYSA-N | |
| 19847174 | |
| C1C(CO1)C(=O)O |
| C4H6O3 | |
| MFCD08669505 | |
| 3-oxetanecarboxylic acid, 3-oxetanecarboxylicacid, acmc-20ahnr, oxetane-3-carboxylc acd, oxetane-3-carboxylicacid | |
| oxetane-3-carboxylic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Store at -20°C; Store under Argon
RUO – Research Use Only