accessibility menu, dialog, popup

UserName

OOOH

CAS RN 114012-41-8

IUPAC Name: oxetane-3-carboxylic acid
Synonyms: oxetane-3-carboxylic acid
Molecular Weight (g/mol): 102.09
Molecular Formula: C4H6O3
InChi Key: UWOTZNQZPLAURK-UHFFFAOYSA-N
SMILES: OC(=O)C1COC1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results