missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Oxelumab Recombinant Human Monoclonal Antibody (R4930 (Oxelumab))
GREENER_CHOICE

Catalog No. PIMA548001
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA548001 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA548001 Supplier Invitrogen™ Supplier No. MA548001
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Human Recombinant Monoclonal Antibody

Specificity: Binds to human OX40L.

Oxelumab is a biosimilar that targets TNFSF4.

Specifications

Antigen Oxelumab
Applications Flow Cytometry, Inhibition Assays
Classification Recombinant Monoclonal
Clone R4930 (Oxelumab)
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% ProClin 300
Gene Alias huMAb OX40L; R4930; RG4930; RO4989991
Host Species Human
Immunogen Oxelumab binds to human OX40L, the ligand of OX40, stimulatory receptor that is expressed on activated T cells, natural killer (NK) cells and natural killer T (NKT) cells.
Purification Method Protein A
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less